Learn More
Potassium hexacyanoferrate(II) trihydrate, ACS, 98.5-102.0%, Thermo Scientific Chemicals
£29.65 - £286.43
Chemical Identifiers
CAS | 14459-95-1 |
---|---|
Molecular Formula | C6H6FeK4N6O3 |
Molecular Weight (g/mol) | 422.39 |
MDL Number | MFCD00167023 |
InChI Key | NLYKFUFIUWPPIZ-UHFFFAOYSA-N |
IUPAC Name | tetrapotassium hexacyanoironbis(ylium) trihydrate |
SMILES | O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337688
|
Thermo Scientific Alfa Aesar
033358.22 |
100 g |
£29.65
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327688
|
Thermo Scientific Alfa Aesar
033358.36 |
500 g |
£86.73
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347688
|
Thermo Scientific Alfa Aesar
033358.A3 |
2 kg |
£286.43
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium hexacyanoferrate(II) trihydrate is widely used in anticaking agent for both road and table salt. It is involved in the purification of tin and separation of copper from molybdenum ores. It plays a vital role in the in production of wine and citric acid. In analytical chemistry, it is used to determine the concentration of potassium permanganate. It is in combination with potassium ferricyanide and phosphate buffered solution acts as buffer for beta-galatosidase. Further, it is used as fertilizers for plants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14459-95-1 | |
422.39 | |
NLYKFUFIUWPPIZ-UHFFFAOYSA-N | |
O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N |
C6H6FeK4N6O3 | |
MFCD00167023 | |
tetrapotassium hexacyanoironbis(ylium) trihydrate |
Specifications
69°C to 71°C | |
Yellow | |
Crystalline | |
MFCD00167023 | |
Soluble in water. Insoluble in ethanol and ether. | |
O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N | |
422.39 | |
98.5 to 102.0% | |
Odorless | |
Potassium hexacyanoferrate(II) trihydrate |
14459-95-1 | |
9.5 | |
C6H6FeK4N6O3 | |
14,7631 | |
NLYKFUFIUWPPIZ-UHFFFAOYSA-N | |
tetrapotassium hexacyanoironbis(ylium) trihydrate | |
422.39 (368.36 Anhydrous) | |
ACS | |
1.85 |
Safety and Handling
missing translation for 'einecsNumber' : 237-722-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Potassium hexacyanoferrate(II) trihydrate
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Potassium ferrocyanide trihydrate
RUO – Research Use Only