missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hexacyanoferrate(II) Trihydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. p-7073423 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 10568630

Brand: Fisher Chemical P/4960/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium Hexacyanoferrate(II) Trihydrate, Certified AR for Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Melting Point 70°C
Color Yellow
pH 9.5
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 422.4g/mol
Packaging Plastic powder jar
Product Identifier
  • Potassium hexacyanoferrate (II)
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : Potassium ferrocyanide trihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.