missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hexacyanoferrate(III), Certified AR for Analysis, Fisher Chemical™

£153.00 - £395.00

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559Show MoreShow Less
PubChem CID 26250
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10050320
SDS View Documents
Fisher Chemical
P/4880/50
250 g
£153.00
250g
Please to purchase this item. Need a web account? Register with us today!
10578440
SDS View Documents
Fisher Chemical
P/4880/53
500 g
£293.00
500g
Please to purchase this item. Need a web account? Register with us today!
10234110
SDS View Documents
Fisher Chemical
P/4880/60
1 kg
£395.00
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Hexacyanoferrate(III), Certified AR for Analysis, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.