missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hexacyanoferrate(Iii), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
500 g
1 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Quantity | 250 g |
Linear Formula | K3Fe(CN)6 |
IUPAC Name | tripotassium;iron(3+);hexacyanide |
Formula Weight | 329.24g/mol |
Percent Purity | ≥99% |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Potassium Hexacyanoferrate(Iii), Reag. Ph. Eur. reag. ISO |
Safety and Handling


Product Identifier
- Potassium hexacyanoferrate(III)
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
- P264-Wash skin thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Tripotassium hexacyanoferrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Hexacyanoferrate(Iii), Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction