missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexafluorophosphate, 98%, Thermo Scientific Chemicals

Product Code. 11439992
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Potassium hexafluorophosphate is used to prepare 1-butyl-3-methylimidazolium hexaflurorphospahte and other organic hexafluorophospahte compounds such as alkylaryliodonium hexafluorophopahtes and triarylsulfonium hexafluorophosphates. It is also used in the cationic polymerization reactions as photo initiators. It involves in producing catalyst for peptide coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hexafluorophosphate is used to prepare 1-butyl-3-methylimidazolium hexaflurorphospahte and other organic hexafluorophospahte compounds such as alkylaryliodonium hexafluorophopahtes and triarylsulfonium hexafluorophosphates. It is also used in the cationic polymerization reactions as photo initiators. It involves in producing catalyst for peptide coupling reactions.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17084-13-8
Molecular Formula F6KP
Molecular Weight (g/mol) 184.062
MDL Number MFCD00011412
InChI Key YZDGRYDIGCWVND-UHFFFAOYSA-N
Synonym potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6
PubChem CID 23688904
IUPAC Name potassium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.[K+]

Specifications

Melting Point 575°C
pH 3.0 to 7.0
Quantity 250 g
UN Number UN3260
Solubility Information Soluble in water.
Formula Weight 184.07
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.55
Chemical Name or Material Potassium hexafluorophosphate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'