Learn More
Potassium hexafluorophosphate, 98%, Thermo Scientific Chemicals
£22.70 - £217.00
Chemical Identifiers
CAS | 17084-13-8 |
---|---|
Molecular Formula | F6KP |
Molecular Weight (g/mol) | 184.062 |
MDL Number | MFCD00011412 |
InChI Key | YZDGRYDIGCWVND-UHFFFAOYSA-N |
Synonym | potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6 |
PubChem CID | 23688904 |
IUPAC Name | potassium;hexafluorophosphate |
SMILES | F[P-](F)(F)(F)(F)F.[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11429992
|
Thermo Scientific Alfa Aesar
A12080.18 |
50 g |
£22.70
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439992
|
Thermo Scientific Alfa Aesar
A12080.30 |
250 g |
£75.20
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419992
|
Thermo Scientific Alfa Aesar
A12080.0B |
1000 g |
£217.00
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium hexafluorophosphate is used to prepare 1-butyl-3-methylimidazolium hexaflurorphospahte and other organic hexafluorophospahte compounds such as alkylaryliodonium hexafluorophopahtes and triarylsulfonium hexafluorophosphates. It is also used in the cationic polymerization reactions as photo initiators. It involves in producing catalyst for peptide coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPotassium hexafluorophosphate is used to prepare 1-butyl-3-methylimidazolium hexaflurorphospahte and other organic hexafluorophospahte compounds such as alkylaryliodonium hexafluorophopahtes and triarylsulfonium hexafluorophosphates. It is also used in the cationic polymerization reactions as photo initiators. It involves in producing catalyst for peptide coupling reactions.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and oxidizing agents.
Chemical Identifiers
17084-13-8 | |
184.062 | |
YZDGRYDIGCWVND-UHFFFAOYSA-N | |
23688904 | |
F[P-](F)(F)(F)(F)F.[K+] |
F6KP | |
MFCD00011412 | |
potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6 | |
potassium;hexafluorophosphate |
Specifications
575°C | |
3.0 to 7.0 | |
MFCD00011412 | |
potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6 | |
YZDGRYDIGCWVND-UHFFFAOYSA-N | |
potassium;hexafluorophosphate | |
23688904 | |
98% | |
Hygroscopic | |
Potassium hexafluorophosphate |
17084-13-8 | |
F6KP | |
UN3260 | |
Soluble in water. | |
F[P-](F)(F)(F)(F)F.[K+] | |
184.062 | |
184.07 | |
Odorless | |
2.55 |
Safety and Handling
GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P301+P330+P331-P302+P352-P304+P340-P305+P351+P338-P310-P312-P362-P501c
H302+H312+H332-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 241-143-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only