missing translation for 'onlineSavingsMsg'
Learn More
Potassium hexafluorotitanate, 97%, Thermo Scientific Chemicals Product Code.: 11414303

Potassium hexafluorotitanate, 97%, Thermo Scientific Chemicals

Product Code. 11414303
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11414303

Brand: Thermo Scientific Alfa Aesar A10915.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium hexafluorotitanate is used as analytical reagent, alloying additive, flame retardant and flux agent for casting. It is also used in the manufacture of titanium acid and aluminum alloys containing titanium. It is also employed in metal surface treatment and finishing, detergent applications and dental impression molding. Further, it is used as a catalyst for polypropylene synthesis. In addition to this, it is used in the extraction-refining-processing of metals, electrical-electronics engineering, textile processing and in ceramics-glass industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hexafluorotitanate is used as analytical reagent, alloying additive, flame retardant and flux agent for casting. It is also used in the manufacture of titanium acid and aluminum alloys containing titanium. It is also employed in metal surface treatment and finishing, detergent applications and dental impression molding. Further, it is used as a catalyst for polypropylene synthesis. In addition to this, it is used in the extraction-refining-processing of metals, electrical-electronics engineering, textile processing and in ceramics-glass industries.

Solubility
Soluble in hot water. Slightly soluble in cold water and inorganic acid. Insoluble in ammonia.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16919-27-0
Molecular Formula F6K2Ti
MDL Number MFCD00011380

Specifications

Boiling Point 235°C to 237°C
Quantity 100 g
Linear Formula K2TiF6
UN Number UN3288
Solubility Information Soluble in hot water. Slightly soluble in cold water and inorganic acid. Insoluble in ammonia.
Formula Weight 240.07
Percent Purity 97%
Chemical Name or Material Potassium hexafluorotitanate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10188443 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberA10915.36,A10915.0E,A10915.22
Lot Number10234226 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10915.36,A10915.0E,A10915.22
Lot Number10160597 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10915.36,A10915.0E,A10915.22
Lot Number10187163 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10915.36,A10915.0E,A10915.22
Lot Number10168050 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10915.36,A10915.0E,A10915.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium hexafluorotitanate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.