Learn More
Potassium hexafluorotitanate, 97%, Thermo Scientific Chemicals
Description
Potassium hexafluorotitanate is used as analytical reagent, alloying additive, flame retardant and flux agent for casting. It is also used in the manufacture of titanium acid and aluminum alloys containing titanium. It is also employed in metal surface treatment and finishing, detergent applications and dental impression molding. Further, it is used as a catalyst for polypropylene synthesis. In addition to this, it is used in the extraction-refining-processing of metals, electrical-electronics engineering, textile processing and in ceramics-glass industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium hexafluorotitanate is used as analytical reagent, alloying additive, flame retardant and flux agent for casting. It is also used in the manufacture of titanium acid and aluminum alloys containing titanium. It is also employed in metal surface treatment and finishing, detergent applications and dental impression molding. Further, it is used as a catalyst for polypropylene synthesis. In addition to this, it is used in the extraction-refining-processing of metals, electrical-electronics engineering, textile processing and in ceramics-glass industries.
Solubility
Soluble in hot water. Slightly soluble in cold water and inorganic acid. Insoluble in ammonia.
Notes
Incompatible with oxidizing agents.

Specifications
Specifications
Boiling Point | 235°C to 237°C |
Quantity | 2500 g |
Linear Formula | K2TiF6 |
UN Number | UN3288 |
Solubility Information | Soluble in hot water. Slightly soluble in cold water and inorganic acid. Insoluble in ammonia. |
Formula Weight | 240.07 |
Percent Purity | 97% |
Chemical Name or Material | Potassium hexafluorotitanate |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10188443 | Certificate of Analysis | 13/10/2022 | A10915.36,A10915.0E,A10915.22 |
10234226 | Certificate of Analysis | 22/11/2021 | A10915.36,A10915.0E,A10915.22 |
10160597 | Certificate of Analysis | 22/11/2021 | A10915.36,A10915.0E,A10915.22 |
10187163 | Certificate of Analysis | 22/11/2021 | A10915.36,A10915.0E,A10915.22 |
10168050 | Certificate of Analysis | 22/11/2021 | A10915.36,A10915.0E,A10915.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.