missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hexahydroxoantimonate(V), Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 100 g |
Linear Formula | KSb(OH)6 |
UN Number | UN1549 |
Formula Weight | 262.9g/mol |
Percent Purity | ≥99% |
Chemical Name or Material | Potassium hexahydroxoantimonate(V) |
Safety and Handling
Product Identifier
- Potassium hexahydroxoantimonate(V)
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : potassium hexahydroxoantimonate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'