missing translation for 'onlineSavingsMsg'
Learn More
Potassium hydrogen carbonate, ACS reagent, Thermo Scientific Chemicals Product Code.: 11914311

Potassium hydrogen carbonate, ACS reagent, Thermo Scientific Chemicals

Product Code. 11914311
100 g
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy

Product Code. 11914311

Brand: Thermo Scientific Acros 446301000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3Show MoreShow Less
PubChem CID 516893
ChEBI CHEBI:81862
SMILES [K+].OC([O-])=O

Specifications

Melting Point 292.0°C
Quantity 100 g
Linear Formula KHCO3
Merck Index 15,773
Solubility Information Solubility in water: 333g/L (20°C). Other solubilities: 600g/L water (60°C), practocally insoluble in alcohol, soluble in potassium carbonate solution
IUPAC Name potassium;hydrogen carbonate
Formula Weight 100.12
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Potassium hydrogen carbonate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465452 Certificate TypeCertificate of Analysis Date27/08/2024 Catalog Number446300025,446301000,446305000
Lot NumberS23J771 Certificate TypeCertificate of Analysis Date12/09/2023 Catalog Number446300025,446301000,446305000
Lot NumberA0453338 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number446300025,446301000,446305000
Lot NumberA0450756 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number446300025,446301000,446305000
Lot NumberA0408457 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number446300025,446301000,446305000
Product Identifier
  • Potassium hydrogen carbonate
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium hydrogen carbonate, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.