missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen fluoride, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10134990 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
25g
500g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10134990

Brand: Thermo Scientific Acros 219235000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-29-9
Molecular Formula HF2K
Molecular Weight (g/mol) 78.1
MDL Number MFCD00011403
Synonym Potassium bifluoride

Specifications

Melting Point 239.0°C
Color White to Gray
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Assay Percent Range 99+%
Linear Formula KHF2
UN Number 1811
Fieser 16,283
Merck Index 15, 7731
Solubility Information Solubility in water: 39g/100mL (20°C). Other solubilities: soluble in diluted acids, insoluble in absolute alcohol, soluble in diluted alcohol
Formula Weight 78.1
Percent Purity ≥99%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Potassium hydrogen fluoride, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium hydrogen fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.