missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen fluoride, 99+% (metals basis), Thermo Scientific Chemicals

Product Code. 11327277
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Potassium hydrogen fluoride is widely used as an etchant in glass and cleaning products. It finds an important application in the synthesis of special optical glass viz. crown and crown flint glass and as a catalyst for polymerization. It is involved in the manufacturing of wood preservatives, soldering agents and brazing. It is also used in the manufacturing of organic and inorganic fluorine compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Electrolyte, flux, catalyst in alkylation of benzene with olefins, and in the synthesis of pure KF
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-29-9
MDL Number MFCD00011403
Synonym Potassium bifluoride

Specifications

Melting Point 239°C (decomposition)
pH 1
Physical Form Crystalline
Quantity 500 g
UN Number UN1811
Merck Index 14,7610
Solubility Information Soluble in water and ethanol.
Formula Weight 78.1
Percent Purity ≥99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.370 g/mL
Assay (metals basis)
Chemical Name or Material Potassium hydrogen fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'