missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen L-tartrate, 98+%

Product Code. 11468136
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
1000 g
5000 g
missing translation for 'unitSize'
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11468136

missing translation for 'mfr': Thermo Scientific Alfa Aesar A15281.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 868-14-4 | C4H5KO6 | 188.18 g/mol

Potassium hydrogen L-tartrate is used for stabilizing egg whites, whipped cream and for anti-caking and thickening. It is utilized for the prevention of crystallization of sugar syrups and reduces the discoloration of boiled vegetables. It is an active component in baking powder. It is used in household items in association with lemon juice or white vinegar to clean metals such as brass, aluminum and copper. Furthermore, it serves as a primary reference standard for a pH buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen L-tartrate is used for stabilizing egg whites, whipped cream and for anti-caking and thickening. It is utilized for the prevention of crystallization of sugar syrups and reduces the discoloration of boiled vegetables. It is an active component in baking powder. It is used in household items in association with lemon juice or white vinegar to clean metals such as brass, aluminum and copper. Furthermore, it serves as a primary reference standard for a pH buffer.

Solubility
Soluble in water and dilute mineral acid. Insoluble in alcohol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868-14-4
Molecular Formula C4H5KO6
Molecular Weight (g/mol) 188.18
MDL Number MFCD00065392,MFCD00071626,MFCD00064206
InChI Key KYKNRZGSIGMXFH-ZVGUSBNCSA-M
Synonym potassium bitartrate, l +-potassium hydrogen tartrate, potassium 3-carboxy-2,3-dihydroxypropanoate, monopotassium tartrate, potassium acid tartrate, kaliumtartrat, potassium hydrogentartrate, 1-potassium hydrogentartrate, ksc657e1t, butanedioic acid, 2,3-dihydroxy-2r,3r-, potassium salt 1:1
IUPAC Name 2,3-dihydroxybutanedioic acid;potassium
SMILES [K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O

Specifications

Melting Point ∼267°C (decomposition)
Density 1.954
Quantity 250 g
Beilstein 6119985
Merck Index 14,7615
Solubility Information Soluble in water and dilute mineral acid. Insoluble in alcohol.
PubChem CID 24193652
Formula Weight 188.18
Percent Purity ≥98%
Chemical Name or Material Potassium hydrogen L-tartrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.