missing translation for 'onlineSavingsMsg'
Learn More
Potassium hydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals Product Code.: 11377327

Potassium hydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals

Product Code. 11377327
5 kg
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11377327

Brand: Thermo Scientific Alfa Aesar 011593.A7

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as a fertilizer, food additive, buffering agent, in molecular biology, biochemistry and chromatography. It is useful to increase the partition coefficients of amino acids in aqueous two-phase systems. It plays an important role for increase in rice grain yield by spraying chemicals including ferric chloride, dipotassium hydrogen phosphate and salicylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-11-4
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
MDL Number MFCD00011383
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Color White
Physical Form Powder
Quantity 5 kg
Assay Percent Range 98.0% min.
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 174.18
Grade ACS
Sensitivity Hygroscopic
Appearance White powder
Chemical Name or Material Potassium hydrogen phosphate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS18K027 Certificate TypeCertificate of Analysis Date30/09/2024 Catalog Number011593.A1,011593.30,011593.A7
Lot NumberU21K048 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog Number011593.A1,011593.30,011593.A7
Lot Number10250996 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog Number011593.A1,011593.30,011593.A7
Lot Number10251481 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog Number011593.A1,011593.30,011593.A7
Lot Number10249110 Certificate TypeCertificate of Analysis Date19/01/2024 Catalog Number011593.A1,011593.30,011593.A7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium hydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.