missing translation for 'onlineSavingsMsg'
Learn More
Potassium hydrogen phthalate, ACS reagent, acidimetric standard, Thermo Scientific Chemicals Product Code.: 10351211

Potassium hydrogen phthalate, ACS reagent, acidimetric standard, Thermo Scientific Chemicals

Product Code. 10351211
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10351211

Brand: Thermo Scientific Acros 424065000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.22
MDL Number MFCD00013070
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Melting Point 295.0°C to 300.0°C
Color White
pH 4.00 to 4.02 (0.05M soln. at 25°C ±0.2°C)
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula 2-(HO2C)C6H4CO2K
Quantity 500 g
Beilstein 09, 791
Merck Index 15, 7733
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: 330g/L water (100°C),slightly soluble in alcohol
Formula Weight 204.22
Percent Purity 99.95 to 100.05% (on dried substance)
Grade ACS Reagent
Insoluble Matter 0.005% max
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Potassium hydrogen phthalate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468088 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number424060025,424061000,424065000
Lot NumberA0468478 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number424060025,424061000,424065000
Lot NumberA0457312 Certificate TypeCertificate of Analysis Date10/11/2023 Catalog Number424060025,424061000,424065000
Lot NumberA0359853 Certificate TypeCertificate of Analysis Date31/05/2023 Catalog Number424060025,424061000,424065000
Lot NumberA0448351 Certificate TypeCertificate of Analysis Date30/12/2022 Catalog Number424060025,424061000,424065000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium hydrogen phthalate, ACS reagent, acidimetric standard, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.