missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide, 0.1N Standardized Solution, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
4 L
20 L
Description
Potassium hydroxide, 0.1N Standardized Solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 1 L |
Solubility Information | Freely soluble in water. Soluble in ethanol. |
IUPAC Name | potassium;hydroxide |
Formula Weight | 56.11 |
Appearance | Colorless liquid |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Chemical Name or Material | Potassium hydroxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'