missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, 50% w/v aq. soln.

Product Code. p-4782081
Click to view available options
Quantity:
500 mL
1 L
4 L
Unit Size:
1 L
4 L
500mL
This item is not returnable. View return policy

Product Code. 11415040

Brand: Thermo Scientific Alfa Aesar 035621.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1310-58-3 | HKO | 56.11 g/mol

Potassium hydroxide acts as the precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
Density 1.5
Solubility Information Miscible with water and ethanol.
Quantity 500 mL
Formula Weight 56.11
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Chemical Name or Material Potassium hydroxide
compliance-icons compliance-icons
Product Identifier
  • Potassium hydroxide, 50% w/v aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Potassium hydroxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.