missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, ≥85%, Honeywell Fluka™
Click to view available options
Quantity:
250 g
500 g
1 kg
2 kg
Descripción
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Especificaciones
Color | White |
Linear Formula | KOH |
UN Number | UN1813 |
Quantity | 250 g |
IUPAC Name | potassium hydroxide |
Formula Weight | 56.11g/mol |
Percent Purity | ≥85% (KOH basis) |
Vapor Pressure | 1 mmHg (719°C) |
Chemical Name or Material | Potassium hydroxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'