missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, ACS, 85% min, K{2}CO{3} 2.0% max

Product Code. 11336808
Click to view available options
Quantity:
50 g
500 g
2 kg
10 kg
Unit Size:
10kg
2kg
500g
50g
This item is not returnable. View return policy

Product Code. 11336808

Brand: Thermo Scientific Alfa Aesar 013451.A3

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use in Soap Manufacture, in Bleaching, As An Electrolyte | CAS: 1310-58-3 | HKO | 56.11 g/mol

In soap manufacture, in bleaching, as an electrolyte, as an absorbent, in dyestuffs, in liquid fertilizer, in electroplating, as a reagentPotassium hydroxide acts as a precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further, it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

Boiling Point 1320°C to 1324°C
Melting Point 360°C
Color White
Physical Form Pellets
Quantity 2 kg
Assay Percent Range 85% min.
UN Number 1813
Solubility Information Soluble in water,alcohol,and glycerol. Slightly soluble in ether.
IUPAC Name potassium hydroxide
Formula Weight 56.11
Grade ACS
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Appearance White pellets
Vapor Pressure 0.13 hPa at 611°C.
Density 2.044 g/cm3
Chemical Name or Material Potassium Hydroxide, For ACS analysis; Usually contains 10-15% water
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.