Learn More
Potassium hydroxide, ACS, 85% min, K{2}CO{3} 2.0% max, Thermo Scientific Chemicals
For Use in Soap Manufacture, in Bleaching, As An Electrolyte
£26.83 - £339.69
Chemical Identifiers
CAS | 1310-58-3 |
---|---|
Molecular Formula | HKO |
Molecular Weight (g/mol) | 56.11 |
MDL Number | MFCD00003553 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
PubChem CID | 14797 |
ChEBI | CHEBI:32035 |
SMILES | [OH-].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316808
|
Thermo Scientific Alfa Aesar
013451.18 |
50 g |
£26.83
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11326808
|
Thermo Scientific Alfa Aesar
013451.36 |
500 g |
£32.66
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336808
|
Thermo Scientific Alfa Aesar
013451.A3 |
2 kg |
£86.93
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346808
|
Thermo Scientific Alfa Aesar
013451.A9 |
10 kg |
£339.69
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
In soap manufacture, in bleaching, as an electrolyte, as an absorbent, in dyestuffs, in liquid fertilizer, in electroplating, as a reagentPotassium hydroxide acts as a precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further, it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
[OH-].[K+] |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 |
Specifications
1320°C to 1324°C | |
1310-58-3 | |
Pellets | |
MFCD00003553 | |
14,7640 | |
Soluble in water,alcohol,and glycerol; Slightly soluble in ether. | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
≥85% | |
Odorless | |
2.044 | |
Potassium hydroxide |
360°C | |
13.5 | |
HKO | |
UN1813 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
14797 | |
56.11 | |
ACS | |
Air sensitive; Hygroscopic | |
K2CO3 2.0% max |
Safety and Handling
GHS H Statement
H314-H302;
Causes severe skin burns and eye damage.
Harmful if swallowed.
P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POTASSIUM HYDROXIDE, SOLID
missing translation for 'einecsNumber' : 215-181-3
missing translation for 'rtecsNumber' : TT2100000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only