missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide, ACS Reagent, pellets, ≥85%, Honeywell Fluka™
Click to view available options
25 kg
Description
Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 1310-58-3 |
Molecular Formula | HKO |
MDL Number | MFCD00003553 |
UN Number | UN1813 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Quantity | 25 kg |
SMILES | [OH-].[K+] |
IUPAC Name | potassium hydroxide |
Molecular Weight (g/mol) | 56.11 |
Chemical Name or Material | Potassium hydroxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'