missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, ca. 50% solution in water

Product Code. 10493165 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
This item is not returnable. View return policy

Product Code. 10493165

Brand: Thermo Scientific Acros 396060025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1310-58-3 | HKO | 56.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1310-58-3, 7732-18-5
Molecular Formula HKO
Linear Formula KOH
MDL Number MFCD00003553
Density 1.3260g/mL
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and glycerol
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Quantity 2.5 kg
SMILES [OH-].[K+]
IUPAC Name potassium hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11
Packaging Plastic bottle
Specific Gravity 1.326
Chemical Name or Material Potassium hydroxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.