missing translation for 'onlineSavingsMsg'
Learn More
Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ Product Code.: 10366240

Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10366240
500 g
Click to view available options
Quantity:
500 g
1 kg
5 kg
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Product Code. 10366240

Brand: Fisher Chemical P/5640/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

HKO, CAS Number-1310-58-3, potash lye, caustic potash, hydroxyde de potassium, caswell no. 693, potassium hydrate, potassium hydroxide, potasse caustique, potassa, potassium hydroxide solution, potassium hydroxide k oh, 500g, 1320 deg.C, CHEBI:32035, White, 56.11g/mol, KWYUFKZDYYNOTN-UHFFFAOYSA-M

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

pH 13.5
Color White
Boiling Point 1320°C
Melting Point 360°C
Quantity 500 g
IUPAC Name potassium;hydroxide
Formula Weight 56.11g/mol
Packaging Plastic powder jar
Physical Form Solid
compliance-icons compliance-icons
Product Identifier
  • Potassium hydroxide
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Potassium hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.