missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide concentrate, For 1L standard solution, 1.0 M KOH (1.0N), Honeywell Fluka™
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Boiling Point | 100°C |
CAS Min % | ≥25.0000%, ≥50.0000% |
CAS Max % | ≤50.0000%, ≤75.0000% |
Linear Formula | KOH |
UN Number | UN1814 |
Density | 1.29 - 1.37 g/cm3 |
Quantity | 6 x 1 Ea. |
Formula Weight | 56.11 |
Packaging | Ampoule |
Physical Form | Concentrate |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'