missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, Extra Pure, SLR, Flake, Fisher Chemical

Product Code. 10428800
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

pH 13.5
Color White
Boiling Point 1320°C
CAS 1310-58-3
Melting Point 360°C
Molecular Formula HKO
MDL Number MFCD00003553
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Quantity 1 kg
SMILES [OH-].[K+]
IUPAC Name potassium hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11g/mol
Packaging Plastic powder jar
Physical Form Solid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium hydroxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.