missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, Pellets, Multi-Compendial, N.F., J.T. Baker™
£153.00 - £375.00
Chemical Identifiers
CAS | 1310-58-3 |
---|---|
Molecular Formula | HKO |
Molecular Weight (g/mol) | 56.11 |
MDL Number | MFCD00003553 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
PubChem CID | 14797 |
ChEBI | CHEBI:32035 |
IUPAC Name | potassium;hydroxide |
SMILES | [OH-].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15508094
|
Avantor J.T.Baker
3152-01 |
500 g | Poly Bottle |
£153.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15518094
|
Avantor J.T.Baker
3152-05 |
2.5 kg | Poly Bottle |
£375.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
potassium;hydroxide |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 | |
[OH-].[K+] |
Specifications
1310-58-3 | |
MFCD00003553 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium;hydroxide | |
14797 | |
56.11 | |
1L = 2.04kg | |
Potassium Hydroxide |
HKO | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
Multi-Compendial/NF | |
Pellet |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.