missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, pure, 8N solution in water, Thermo Scientific Chemicals

Product Code. 10472463
Click to view available options
Quantity:
500 mL
2 L
10 L
Packaging:
Plastic bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3, 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 2 L
Linear Formula KOH
Merck Index 15,7761
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and glycerol
IUPAC Name potassium hydroxide
Formula Weight 56.11
Grade Pure
Concentration or Composition (by Analyte or Components) 7.992 to 8.008N (20°C)
Packaging Plastic bottle
Specific Gravity 1.326
Density 1.3260g/mL
Chemical Name or Material Potassium hydroxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium hydroxide, 30-50% solution in water
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Potassium hydroxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'