missing translation for 'onlineSavingsMsg'
Learn More
Learn More

- Home
- Products
- Chemicals
- Bases
- Potassium Hydroxide
- Potassium hydroxide, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 85-100.5%, pellets, Honeywell Fluka™
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Potassium hydroxide, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 85-100.5%, pellets, Honeywell Fluka™
Product Code. 15600810
Shop All Honeywell Fluka Products

25 kg
Click to view available options
Quantity:
1 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
This item is not returnable.
View return policy
Product Code. 15600810
Brand: Honeywell-Fluka 0600525KG
This item is not returnable.
View return policy
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Chemical Identifiers
CAS | 1310-58-3 |
Molecular Formula | HKO |
Molecular Weight (g/mol) | 56.11 |
MDL Number | MFCD00003553 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693Show MoreShow Less |
PubChem CID | 14797 |
ChEBI | CHEBI:32035 |
SMILES | [OH-].[K+] |
Specifications
CAS Max % | 100.0000% |
Assay Percent Range | 85 to 100.5% |
Linear Formula | KOH |
UN Number | UN1813 |
Quantity | 25 kg |
IUPAC Name | potassium;hydroxide |
Formula Weight | 56.11 |
Percent Purity | 85 to 100.5% |
Grade | puriss. |
Packaging | Paper / Poly Bag |
Vapor Pressure | 1mmHg (719°C) |
Physical Form | Pellet |
Chemical Name or Material | Potassium hydroxide, Puriss., meets analytical specification of BP, NF, Ph. Eur., 85-100.5%, pellets |
Show More Show Less |
Product Suggestions
You may be interested in these alternatives
-
Product Code. 10454015
-
Product Code. 15623070
-
Product Code. 15610810
-
Product Code. 15620810
-
Product Code. 15630810
-
Product Code. 13428578
Customers who viewed this item also viewed. This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-5 of 12
-
Catalog No. 10316380
£75.90 / 2.50 L -
Catalog No. 10596814
£145.00 / 50mL -
Catalog No. 10304980
£97.20 / 2.50 L -
Catalog No. 10703811
£258.00 / 500g -
Catalog No. 10793611
£101.00 / 1kg -
Catalog No. 10112640
£50.40 / 1kg -
Catalog No. 10401841
£22.81 / 500mL -
Catalog No. 10117113
£25.71 / 100g -
Catalog No. 15681810
£34.55 / 25g -
Catalog No. 10448990
£67.35 / 1kg -
Catalog No. 10509390
£41.35 / 500g -
Catalog No. 10113190
£46.46 / 1kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium hydroxide, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 85-100.5%, pellets, Honeywell Fluka™ >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Spot an opportunity for improvement?Share a Content Correction