missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, Puriss. p.a., Reag. Ph. Eur., ≥85%, pellets, Honeywell Fluka™

Puriss. p.a., Reag. Ph. Eur.,≥85%, pellets

£36.75 - £702.03

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]
View More Specs

Products 7
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15633070
View Documents Promotion Details
Honeywell-Fluka
30603-500G
500 g
£36.75
500g
Please to purchase this item. Need a web account? Register with us today!
15693060
View Documents Promotion Details
Honeywell-Fluka
30603-1KG
1 kg
£50.98
1kg
Please to purchase this item. Need a web account? Register with us today!
15603070
View Documents Promotion Details
Honeywell-Fluka
30603-2.5KG
2.5 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15663070
View Documents Promotion Details
Honeywell-Fluka
30603-6X500G
6 x 500 g
£134.00
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15643070
View Documents Promotion Details
Honeywell-Fluka
30603-5KG
5 kg
£203.27
5kg
Please to purchase this item. Need a web account? Register with us today!
15653070
View Documents Promotion Details
Honeywell-Fluka
30603-6X1KG
6 x 1 kg
£265.04
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15623070
View Documents Promotion Details
Honeywell-Fluka
30603-4X5KG
4 x 5 kg
£702.03
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
[OH-].[K+]
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035

Specifications

1310-58-3
KOH
1813
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium;hydroxide
14797
56.11g/mol
puriss. p.a.
Potassium Hydroxide, Reag. Ph. Eur.
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
[OH-].[K+]
56.11
CHEBI:32035
≥85%
1mmHg (719°C)
Videos
Safety and Handling

Safety and Handling

P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

H290-H302-H314

missing translation for 'einecsNumber' : 215-181-3

missing translation for 'rtecsNumber' : TT2100000

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications