missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, Semiconductor grade, Honeywell
Click to view available options
Quantity:
100 g
500 g
Specifications
Linear Formula | KOH |
UN Number | 1813 |
Quantity | 100 g |
Formula Weight | 56.11g/mol |
Percent Purity | 99.99% (trace metals basis) |
Grade | Semiconductor |
Vapor Pressure | 1mmHg (719°C) |
Chemical Name or Material | Potassium Hydroxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'