missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide solution, 0.1M, Honeywell Fluka™
54.87 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
1 L
10 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Linear Formula | KOH |
Concentration | 0.1M |
UN Number | NONH for all modes of transport |
Quantity | 1 L |
Formula Weight | 56.11g/mol |
Chemical Name or Material | Potassium hydroxide solution |
Safety and Handling
Product Identifier
- Potassium hydroxide solution
- Warning
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : potassium hydroxide;caustic potash
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'