missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide Solution, 0.1M in methanol, Honeywell Fluka™
Click to view available options
Quantity:
500 mL
2 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
Linear Formula | KOH |
Concentration | 0.1M in methanol |
UN Number | 1230 |
Density | 0.81g/mL at 25°C |
Quantity | 2 L |
IUPAC Name | potassium;hydroxide |
Flash Point | 11°C (51.8°F) |
Formula Weight | 56.11g/mol |
Chemical Name or Material | Potassium Hydroxide Solution |
Safety and Handling
Product Identifier
- Potassium hydroxide solution
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : methanol, potassiumhydroxide
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'