missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution 1M (1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical

Product Code. 10736292 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 Ampule
Unit Size:
1 ampoule
This item is not returnable. View return policy

Product Code. 10736292

Brand: Fisher Chemical J/6630C/05

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For volumetric analysis
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
MDL Number 3553
Quantity 1 Ampule
Packaging Ampoule
Physical Form Liquid
compliance-icons
Product Identifier
  • Potassium hydroxide, 1N solution in water
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.