missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Potassium Hydroxide Solution

Brand:  Lovibond™ 2418634

Code : NEW

Additional Details : CAS Number : 1310-58-3 Weight : 0.08700kg Transport : UN number : 1814 Chem class : 8 Pack group : II


Product Code. 10029811

  • £14.40 / 50mL
Estimated Shipment: 24-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Product Type: Potassium Hydroxide Solution

Specifications

Specifications

Potassium Hydroxide Solution 45%
BOD
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • potassium hydroxide solution 45%

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4

Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P390-Absorb spillage to prevent material damage.

Supplemental information
  • MIXTURE LIST-Contains : potassium hydroxide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers