missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide Solution, TraceSELECT™, ≥30% (in water), Honeywell™ Fluka™
Click to view available options
Quantity:
250 mL
Packaging:
Poly bottle
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | 14 (at 20°C) |
Color | Colorless |
Boiling Point | >100°C |
CAS | 1310-58-3 |
Molecular Formula | HKO |
Linear Formula | KOH |
MDL Number | MFCD00003553 |
UN Number | UN1814 |
Density | 1.29g/mL at 20°C |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'