missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate, Honeywell Fluka™

Product Code. 15682990
Click to view available options
Quantity:
100 g
250 g
1 kg
25 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., 99.7-100.4%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-05-6
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

Quantity 100 g
Linear Formula KIO3
UN Number 1479
Formula Weight 214g/mol
Percent Purity 99.7 to 104%
Grade puriss. p.a.
Density 3.93g/mL at 25°C
Chemical Name or Material Potassium Iodate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.