Learn More
Potassium iodide, 99.995% (metals basis), Thermo Scientific Chemicals
£47.60 - £414.00
Chemical Identifiers
CAS | 7681-11-0 |
---|---|
Molecular Formula | IK |
Molecular Weight (g/mol) | 166.003 |
MDL Number | MFCD00011405 |
InChI Key | NLKNQRATVPKPDG-UHFFFAOYSA-M |
Synonym | potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide |
PubChem CID | 4875 |
ChEBI | CHEBI:8346 |
IUPAC Name | potassium;iodide |
SMILES | [K+].[I-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11331568
|
Thermo Scientific Alfa Aesar
010978.06 |
5 g |
£47.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11321568
|
Thermo Scientific Alfa Aesar
010978.14 |
25 g |
£119.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11341568
|
Thermo Scientific Alfa Aesar
010978.22 |
100 g |
£414.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium iodide is used with silver nitrate to make silver iodide an important chemical in film photography. Medicinally it is used for thyroid blockade, thyroid storm and also as an expectorant. It is a component in some hair treatment chemicals. It is also used as a fluorescence quenching agent in biomedical research, an application that takes advantage of collisional quenching of fluorescent substances by the iodide ion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7681-11-0 | |
166.003 | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
4875 | |
potassium;iodide |
IK | |
MFCD00011405 | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
CHEBI:8346 | |
[K+].[I-] |
Specifications
1330°C | |
7681-11-0 | |
Crystalline | |
IK | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
potassium;iodide | |
4875 | |
166.01 | |
1.677 | |
White crystalline powder | |
3.12 |
681°C | |
White | |
99.995% (metals basis) | |
MFCD00011405 | |
Soluble in water,alcohol,methanol,acetone and ethanol. | |
[K+].[I-] | |
166.003 | |
CHEBI:8346 | |
Odorless | |
Hygroscopic | |
1.33 hPa (1mm Hg) at 745°C | |
Potassium iodide |
Safety and Handling
P260-P264b-P270-P314-P501c
H372
missing translation for 'einecsNumber' : 213-659-4
missing translation for 'rtecsNumber' : TT2975000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only