missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, 99.995%, (trace metal basis), extra pure, Thermo Scientific Chemicals

£36.28 - £311.55

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10557183
View Documents Promotion Details
Thermo Scientific Acros
193790100
10 g Glass bottle
£36.28
10g
Please to purchase this item. Need a web account? Register with us today!
10562981
View Documents Promotion Details
Thermo Scientific Acros
193790500
50 g Glass bottle
£73.77
50g
Please to purchase this item. Need a web account? Register with us today!
10338753
View Documents Promotion Details
Thermo Scientific Acros
193792500
250 g Glass bottle
£311.55
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

7681-11-0
166
NLKNQRATVPKPDG-UHFFFAOYSA-M
4875
potassium;iodide
IK
MFCD00011405
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
CHEBI:8346
[K+].[I-]

Specifications

1330.0°C
7681-11-0
Crystals or Powder
IK
MFCD00011405
15, 7764
Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol
[K+].[I-]
166
CHEBI:8346
99.995%
Potassium iodide
680.0°C
White
99.995%
KI
06,227; 11,440
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium;iodide
4875
166
Trace Metal Basis
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 231-659-4

Product Identifier
  • Potassium iodide

Signal Word
  • Danger

Hazard Category
  • Specific target organ toxicity Category 1

Hazard Statement
  • H372-Causes damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : Potassium iodide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only