missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium iodide, 99%, extra pure, briquettes, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1330.0°C |
Melting Point | 680.0°C |
Color | White |
Physical Form | Briquettes |
Quantity | 2.5 kg |
Assay Percent Range | 98.5% min. (on dried substance) (Argentometry) |
Linear Formula | KI |
Fieser | 06,227; 11,44 |
Merck Index | 15,7764 |
Solubility Information | Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466123 | Certificate of Analysis | 20/09/2024 | 196730025,196731000,196730100,196735000 |
A0459801 | Certificate of Analysis | 11/06/2024 | 196730025,196731000,196730100,196735000 |
A0462331 | Certificate of Analysis | 09/05/2024 | 196730025,196731000,196730100,196735000 |
A0444289 | Certificate of Analysis | 21/09/2023 | 196730025,196731000,196730100,196735000 |
A0443016 | Certificate of Analysis | 25/08/2023 | 196730025,196731000,196730100,196735000 |
Safety and Handling

Product Identifier
- Potassium iodide
- Danger
- Specific target organ toxicity Category 1
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium iodide, 99%, extra pure, briquettes, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction