missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, 99%, extra pure, briquettes, Thermo Scientific Chemicals

Product Code. 11964211
10 kg, Plastic drum
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodideShow MoreShow Less
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330.0°C
Melting Point 680.0°C
Color White
Physical Form Briquettes
Quantity 10 kg
Assay Percent Range 98.5% min. (on dried substance) (Argentometry)
Linear Formula KI
Fieser 06,227; 11,44
Merck Index 15,7764
Solubility Information Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol
Formula Weight 166
Percent Purity 99%
Grade Extra Pure
Loss on Drying 1% max.
Packaging Plastic drum
Chemical Name or Material Potassium iodide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466123 Certificate TypeCertificate of Analysis Date20/09/2024 Catalog Number196730025,196731000,196730100,196735000
Lot NumberA0459801 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number196730025,196731000,196730100,196735000
Lot NumberA0462331 Certificate TypeCertificate of Analysis Date09/05/2024 Catalog Number196730025,196731000,196730100,196735000
Lot NumberA0444289 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number196730025,196731000,196730100,196735000
Lot NumberA0443016 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number196730025,196731000,196730100,196735000
compliance-icons
Product Identifier
  • Potassium iodide
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium iodide, 99%, extra pure, briquettes, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.