Learn More
Potassium iodide, 99%, Thermo Scientific Chemicals
Description
Potassium iodide is used in pharmaceutical formulations and in film photography as a source of iodine. It is widely used as an analytical Reagent in iodometric tritrations. Apart from its application as a source of iodide to produce organo iodine compounds such as in Sandmeyer reactions, potassium iodide finds extensive use as a nucleophilic catalyst for several alkylation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium iodide is used in pharmaceutical formulations and in film photography as a source of iodine. It is widely used as an analytical Reagent in iodometric tritrations. Apart from its application as a source of iodide to produce organo iodine compounds such as in Sandmeyer reactions, potassium iodide finds extensive use as a nucleophilic catalyst for several alkylation reactions.
Solubility
Soluble in water
Notes
Being a mild reducing agent, it gets converted into iodine by oxidizing reagents. Potassium iodide should be handled with gloves since it is a mild irritant.

Specifications
Specifications
Boiling Point | 1330°C |
Melting Point | 681°C |
Quantity | 50 g |
Merck Index | 14,7643 |
Solubility Information | Soluble in water |
Formula Weight | 166.01 |
Percent Purity | 99% |
Odor | Odorless |
Refractive Index | 1.677 |
Sensitivity | Air and light sensitive |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10252492 | Certificate of Analysis | 24/09/2024 | A12704.18,A12704.0I,A12704.30,A12704.0B |
10252491 | Certificate of Analysis | 24/09/2024 | A12704.18,A12704.0I,A12704.30,A12704.0B |
10250093 | Certificate of Analysis | 04/04/2024 | A12704.18,A12704.0I,A12704.30,A12704.0B |
10248384 | Certificate of Analysis | 06/11/2023 | A12704.18,A12704.0I,A12704.30,A12704.0B |
10246673 | Certificate of Analysis | 19/05/2023 | A12704.18,A12704.0I,A12704.30,A12704.0B |
Safety and Handling

- Potassium iodide
- Danger
- Specific target organ toxicity Category 1
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.
- MIXTURE LIST-Contains : potassium iodide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.