missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, Extra Pure, SLR, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
250g
500g
5kg
Description
Potassium Iodide, Extra Pure, SLR
Specifications
Specifications
| Boiling Point | 1330°C |
| Melting Point | 680°C |
| Color | White |
| pH | 6 to 8 |
| Physical Form | Solid |
| Quantity | 5 kg |
| Formula Weight | 166g/mol |
| Packaging | Plastic powder jar |
| Vapor Pressure | 1mmHg 745 |
Safety and Handling
Product Identifier
- Potassium iodide
- Danger
- Specific target organ toxicity Category 1
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction