missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium iodide, Puriss., meets analytical spec. of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance), Honeywell Fluka™
Shop All Honeywell Fluka Products

Click to view available options
Quantity:
250 g
1 kg
5 kg
6 x 250 g
25 kg
50 kg
6 x 1 kg
Unit Size:
1kg
250g
25kg
50kg
5kg
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Boiling Point | 1,323°C |
| CAS Max % | 100.0000% |
| Quantity | 6 x 1 kg |
| Assay Percent Range | 99.0 to 100.5% |
| Linear Formula | KI |
| Formula Weight | 166 |
| Grade | puriss. |
| Loss on Drying | max. 0.5% |
| Packaging | Poly Bottle |
| Density | 3.12 g/cm3 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction