missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, ReagentPlus™, Honeywell™
Reagent Grade, 99%
£30.75 - £152.68
Chemical Identifiers
CAS | 7681-11-0 |
---|---|
Molecular Formula | IK |
Molecular Weight (g/mol) | 166.003 |
MDL Number | MFCD00011405 |
InChI Key | NLKNQRATVPKPDG-UHFFFAOYSA-M |
Synonym | potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide |
PubChem CID | 4875 |
ChEBI | CHEBI:8346 |
IUPAC Name | potassium;iodide |
SMILES | [K+].[I-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15691800
|
Honeywell Research Chemicals
207969-100G |
100 g | N/A | ||||||
15601810
|
Honeywell Research Chemicals
207969-500G |
500 g |
£152.68
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7681-11-0 | |
166.003 | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
4875 | |
potassium;iodide |
IK | |
MFCD00011405 | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
CHEBI:8346 | |
[K+].[I-] |
Specifications
7681-11-0 | |
KI | |
NONH for all modes of transport | |
Soluble in water | |
[K+].[I-] | |
166.003 | |
CHEBI:8346 | |
99% | |
1mmHg (745°C) |
IK | |
MFCD00011405 | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
potassium;iodide | |
4875 | |
166g/mol | |
ReagentPlus™ | |
Potassium Iodide |
Safety and Handling
P305 + P351 + P338
H302-H315-H319
missing translation for 'einecsNumber' : 231-659-4
missing translation for 'rtecsNumber' : TT2975000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title