missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide Solution, Ph. Eur., Reagecon™
BIOPHARMA

Product Code. 11454729
Click to view available options
Quantity:
1 L
Packaging:
1L
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 7681-11-0, 7732-18-5
Quantity 1 L
Molecular Formula IK
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES [K+].[I-]
IUPAC Name potassium iodide
Molecular Weight (g/mol) 166.00
PubChem CID 4875
ChEBI CHEBI:8346
Grade Ph. Eur.
Packaging 1L
Chemical Name or Material Potassium Iodide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'