missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide (White, Free-Flowing Granules), Fisher BioReagents
Brand: Fisher BioReagents BP367-500
94.00 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Poly Bottle; 500g
This potassium salt is used in buffers for molecular biology and cell culture applications.Specifications
1330°C | |
680°C | |
White | |
Solid | |
≥99.0 % | |
15, 7764 | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
potassium;iodide | |
4875 | |
166 | |
Pass Test | |
Poly Bottle |
Potassium Iodide | |
7681-11-0 | |
6 to 8 | |
500 g | |
IK | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
[K+].[I-] | |
166.003 | |
CHEBI:8346 | |
≥99.0% | |
0.2% max. | |
1mmHg at 745°C |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Potassium iodide
Signal Word
- Danger
Hazard Category
- Specific target organ toxicity Category 1
Hazard Statement
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.