missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodobismuthate Solution, EP, Reagecon
BIOPHARMA

Product Code. 11464729 Shop All Reagecon Products
Click to view available options
Quantity:
100 mL
Packaging:
Amber Glass Winchester Bottle
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 11464729

Brand: Reagecon 1070600

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured according to the standards of the European Pharmacopoeia (EP) Chapter 4 (4.1.1). Reagecon Potassium Iodobismuthate Solution, EP is a high purity, stable product developed and tested for EP applications.

Reagecon Potassium Iodobismuthate Solution is a high purity, stable product developed and tested for European Pharmacopoeia (EP) applications, including identification reactions of ions and functional groups (2.3.1) and as a reagent in the performance of tests outlined in several EP monographs.

  • This Reagecon EP reagent is accurate, stable and reproducible
  • Stability is proven (either opened or unopened) for the shelf life of the product
  • Packaged in an amber glass bottle for protection from light

Specifications

CAS 41944-01-8
Color Reddish-Brown
Physical Form Liquid
Quantity 100 mL
Molecular Formula BiI7K4
UN Number 2790
Molecular Weight (g/mol) 1253.7
Grade Eur Ph.
Packaging Amber Glass Winchester Bottle
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contains : acetic acid, BismuthSubnitrate Bismuthhydroxide nitrate oxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.