Learn More
Potassium metabisulfite, 97%, extra pure, Thermo Scientific Chemicals
£16.50 - £73.70
Chemical Identifiers
CAS | 16731-55-8 |
---|---|
Molecular Formula | K2O5S2 |
Molecular Weight (g/mol) | 222.31 |
MDL Number | MFCD00167605 |
InChI Key | RWPGFSMJFRPDDP-UHFFFAOYSA-L |
Synonym | potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5 |
PubChem CID | 28019 |
IUPAC Name | dipotassium sulfinatosulfonate |
SMILES | [K+].[K+].[O-]S(=O)S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10722792
|
Thermo Scientific Acros
418270250 |
25 g | Glass bottle |
£16.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10229560
|
Thermo Scientific Acros
418275000 |
500 g | Glass bottle |
£24.70
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10320354
|
Thermo Scientific Acros
418270025 |
2.5 kg | Plastic bottle |
£73.70
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
16731-55-8 | |
222.31 | |
RWPGFSMJFRPDDP-UHFFFAOYSA-L | |
28019 | |
[K+].[K+].[O-]S(=O)S([O-])(=O)=O |
K2O5S2 | |
MFCD00167605 | |
potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5 | |
dipotassium sulfinatosulfonate |
Specifications
150.0°C | |
White to Yellow | |
96% min. (Iodimetry) | |
K2S2O5 | |
15, 7766 | |
Solubility in water: 45g/L (20°C). Other solubilities: insoluble in ethanol | |
[K+].[K+].[O-]S(=O)S([O-])(=O)=O | |
222.31 | |
222.33 | |
Extra Pure |
16731-55-8 | |
Fine Crystalline Powder | |
K2O5S2 | |
MFCD00167605 | |
potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5 | |
RWPGFSMJFRPDDP-UHFFFAOYSA-L | |
dipotassium sulfinatosulfonate | |
28019 | |
97% | |
Potassium metabisulfite, 97% |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Contact with acids liberates toxic gas.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 240-795-3
missing translation for 'rtecsNumber' : TT4920000
missing translation for 'tsca' : TSCA
- Potassium metabisulfite
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
- EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only