missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Metal In Liquid Paraffin, Pure, Fisher Chemical™
£45.85
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11498752
|
Fisher Chemical
P/3720/45 |
10 g | N/A | ||||||
Description
Chemical Identifiers
7740-09-7 | |
K | |
133776 |
Specifications
770°C | |
7740-09-7 | |
Solid | |
133776 | |
Plastic container |
63°C | |
Gray | |
K | |
39.1g/mol | |
133mbar 171 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Potassium (metal) in liquid paraffin
Signal Word
- Danger
Hazard Category
- Substance which\, in contact with water\, emits flammable gases Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.