missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Methoxide, 0.1N, Reagecon™
Click to view available options
Quantity:
500 mL
Specifications
Specifications
CAS | 865-33-8 |
Molecular Formula | CH3KO |
Quantity | 500 mL |
Synonym | potassium methoxide, potassium methylate, potassium methanolate, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok, rntqddwhl@ |
InChI Key | BDAWXSQJJCIFIK-UHFFFAOYSA-N |
Molecular Weight (g/mol) | 70.13 |
PubChem CID | 23664618 |
Safety and Handling
Product Identifier
- 0.1N POTASSIUM METHOXIDE IN METHANOL
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : METHANOL
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'